4-[(2-Bromo-4-fluorophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050791-500MG 050791-2.5G
101893-060EA 364.62 USD
101893-060 101893-062
4-[(2-Bromo-4-fluorophenoxy)methyl]piperidine hydrochloride
4-[(2-Bromo-4-fluorophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆BrClFNO
MDL Number: MFCD13561174
CAS Number: 1220019-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR