3-(2-Bromo-4-fluorophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050788-500MG 050788-2.5G
101893-048EA 364.62 USD
101893-048 101893-050
3-(2-Bromo-4-fluorophenoxy)piperidine hydrochloride
3-(2-Bromo-4-fluorophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄BrClFNO
MDL Number: MFCD13561171
CAS Number: 1220034-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR