3-(4-Cyanophenyl)acrylic acid ≥95%

Supplier: Matrix Scientific

047690-1G 047690-5G
101880-992EA 35.38 USD
101880-992 101880-994
3-(4-Cyanophenyl)acrylic acid ≥95%
3-(4-Cyanophenyl)acrylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₂
MDL Number: MFCD00016845
CAS Number: 18664-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR