5-Nitro-2-(1-pyrrolidinyl)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047682-500MG 047682-1G
101880-970EA 124.62 USD
101880-970 101880-972
5-Nitro-2-(1-pyrrolidinyl)benzenecarbaldehyde ≥95%
5-Nitro-2-(1-pyrrolidinyl)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
MDL Number: MFCD00455053
CAS Number: 30742-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR