3-{4-[(3,4-Dichlorobenzyl)oxy]-3-nitrophenyl}acrylic acid ≥95%
Supplier: Matrix Scientific
047681-500MG
047681-1G
101880-966EA
181.54
USD
101880-966
101880-968
3-{4-[(3,4-Dichlorobenzyl)oxy]-3-nitrophenyl}acrylic acid ≥95%
3-{4-[(3,4-Dichlorobenzyl)oxy]-3-nitrophenyl}acrylic acid
Hazard: Irritant
Formula:
C₁₆H₁₁Cl₂NO₅ |
MDL Number:
MFCD07021303 CAS Number: 885267-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...