2-[(4-Fluorophenyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047678-500MG 047678-1G
101880-954EA 181.54 USD
101880-954 101880-956
2-[(4-Fluorophenyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%
2-[(4-Fluorophenyl)sulfanyl]-5-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₈FNO₃S
MDL Number: MFCD07021301
CAS Number: 35985-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR