4-[(2-Furylmethyl)sulfanyl]-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047677-500MG 047677-1G
101880-950EA 92.31 USD
101880-950 101880-952
4-[(2-Furylmethyl)sulfanyl]-3-nitrobenzenecarbaldehyde ≥95%
4-[(2-Furylmethyl)sulfanyl]-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₂H₉NO₄S
MDL Number: MFCD00173717
CAS Number: 175278-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR