N-(3,4-Dimethylphenyl)-N'-[hydrazino(imino)methyl]guanidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: N'-Amino-N-(3,4-dimethylphenyl)biguanidine hydrochloride

047673-500MG 047673-1G
101880-936EA 181.54 USD
101880-936 101880-938
N-(3,4-Dimethylphenyl)-N'-[hydrazino(imino)methyl]guanidine hydrochloride ≥95%
N-(3,4-Dimethylphenyl)-N'-[hydrazino(imino)methyl]guanidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₇ClN₆
MW: 256.74 g/mol
MDL Number: MFCD09027118
CAS Number: 1171647-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR