4-(4-Fluorophenyl)-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulfide ≥95%

Supplier: Matrix Scientific

047666-500MG 047666-1G
101880-912EA 181.54 USD
101880-912 101880-914
4-(4-Fluorophenyl)-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulfide ≥95%
4-(4-Fluorophenyl)-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulfide

Hazard: Irritant

Formula: C₁₄H₁₀FN₃S
MDL Number: MFCD02720697
CAS Number: 120873-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR