3-Benzylrhodanine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Benzyl-2-thioxo-4-thiazolidinone, 3-Benzyl-2-thioxothiazolidin-4-one
047652-500MG
047652-1G
101880-882EA
181.54
USD
101880-882
101880-884
3-Benzylrhodanine ≥95%
3-Benzylrhodanine
Hazard: Irritant
Formula:
C₁₀H₉NOS₂ MW: 223.31 g/mol |
MDL Number:
MFCD00086924 CAS Number: 10574-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...