N-(4-Fluorophenyl)rhodanine ≥95%
Supplier: Matrix Scientific
047651-500MG
047651-1G
101880-878EA
181.54
USD
101880-878
101880-880
N-(4-Fluorophenyl)rhodanine ≥95%
N-(4-Fluorophenyl)rhodanine
Hazard: Irritant
Formula:
C₉H₆FNOS₂ |
CAS Number:
387-27-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...