3-Methoxy-4-[(4-nitrobenzyl)oxy]benzaldehyde ≥95%

Supplier: Matrix Scientific

047645-500MG 047645-1G
101880-860EA 181.54 USD
101880-860 101880-862
3-Methoxy-4-[(4-nitrobenzyl)oxy]benzaldehyde ≥95%
3-Methoxy-4-[(4-nitrobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₃NO₅
MDL Number: MFCD00016711
CAS Number: 81307-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR