3-(4-Hydroxyphenyl)-1-(4-methoxyphenyl)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-4'-methoxychalcone

047643-500MG 047643-1G
101880-850EA 181.54 USD
101880-850 101880-852
3-(4-Hydroxyphenyl)-1-(4-methoxyphenyl)-2-propen-1-one ≥95%
3-(4-Hydroxyphenyl)-1-(4-methoxyphenyl)-2-propen-1-one

Hazard: Irritant

Formula: C₁₆H₁₄O₃
MW: 254.29 g/mol
MDL Number: MFCD00210515
CAS Number: 69704-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR