3-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}acrylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[(3,4-Dichlorobenzyl)oxy]cinnamic acid

047638-500MG 047638-1G 047638-5G
101880-828EA 278.46 USD
101880-828 101880-830 101880-832
3-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}acrylic acid ≥95%
3-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}acrylic acid

Hazard: Irritant

Formula: C₁₆H₁₂Cl₂O₃
MW: 323.18 g/mol
MDL Number: MFCD00173654
CAS Number: 175136-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR