3',5'-Bis(trifluoromethyl)-2-chloroacetanilide ≥95%
Supplier: Matrix Scientific
047623-500MG
047623-1G
101880-818EA
38.46
USD
101880-818
101880-820
3',5'-Bis(trifluoromethyl)-2-chloroacetanilide ≥95%
3',5'-Bis(trifluoromethyl)-2-chloroacetanilide
Hazard: Irritant
Formula:
C₁₀H₆ClF₆NO |
MDL Number:
MFCD00077474 CAS Number: 790-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...