3-(6-Bromo-1,3-benzodioxol-5-yl)acrylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(6-Bromobenzo[d][1,3]dioxol-5-yl)acrylic acid

047622-500MG 047622-1G
101880-814EA 181.54 USD
101880-814 101880-816
3-(6-Bromo-1,3-benzodioxol-5-yl)acrylic acid ≥95%
3-(6-Bromo-1,3-benzodioxol-5-yl)acrylic acid

Hazard: Irritant

Formula: C₁₀H₇BrO₄
MW: 271.07 g/mol
MDL Number: MFCD00017599
CAS Number: 27452-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR