3-(2-Chlorophenyl)-1-(4-hydroxyphenyl)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4'-hydroxychalcone

047615-500MG 047615-1G
101880-786EA 181.54 USD
101880-786 101880-788
3-(2-Chlorophenyl)-1-(4-hydroxyphenyl)-2-propen-1-one ≥95%
3-(2-Chlorophenyl)-1-(4-hydroxyphenyl)-2-propen-1-one

Hazard: Irritant

Formula: C₁₅H₁₁ClO₂
MW: 258.71 g/mol
Melting Pt: 170 °C
MDL Number: MFCD03788397
CAS Number: 5424-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR