4-[(3,4-Dichlorobenzyl)oxy]-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047611-500MG 047611-1G
101880-774EA 181.54 USD
101880-774 101880-776
4-[(3,4-Dichlorobenzyl)oxy]-3-nitrobenzenecarbaldehyde ≥95%
4-[(3,4-Dichlorobenzyl)oxy]-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₉Cl₂NO₄
MDL Number: MFCD00173646
CAS Number: 175136-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR