4-[(4-Fluorobenzyl)oxy]-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047609-500MG 047609-1G
101880-770EA 181.54 USD
101880-770 101880-772
4-[(4-Fluorobenzyl)oxy]-3-nitrobenzenecarbaldehyde ≥95%
4-[(4-Fluorobenzyl)oxy]-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀FNO₄
MDL Number: MFCD00173649
CAS Number: 175136-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR