Methyl-5-iodsalicylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-hydroxy-5-iodobenzoate

047604-500MG 047604-1G
101880-748EA 181.54 USD
101880-748 101880-750
Methyl-5-iodsalicylate ≥95%
Methyl 5-iodsalicylate

Hazard: Irritant

Formula: C₈H₇IO₃
MW: 278.05 g/mol
MDL Number: MFCD00016462
CAS Number: 4068-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR