3-[1,1'-Biphenyl]-4-ylacrylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Phenylcinnamic acid

047591-500MG 047591-1G 047591-5G
101880-698EA 181.54 USD
101880-698 101880-700 101880-702
3-[1,1'-Biphenyl]-4-ylacrylic acid ≥95%
3-[1,1'-Biphenyl]-4-ylacrylic acid

Hazard: Irritant

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
MDL Number: MFCD00014010
CAS Number: 13026-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR