2-Chloro-N-(5-fluoro-2-methylphenyl)acetamide ≥95%

Supplier: Matrix Scientific

Synonyms: N-(Chloroacetyl)-5-fluoro-2-methylaniline

047581-500MG 047581-1G
101880-656EA 181.54 USD
101880-656 101880-658
2-Chloro-N-(5-fluoro-2-methylphenyl)acetamide ≥95%
2-Chloro-N-(5-fluoro-2-methylphenyl)acetamide

Hazard: Irritant

Formula: C₉H₉ClFNO
MW: 201.63 g/mol
MDL Number: MFCD03788386
CAS Number: 85817-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR