4-[(4-Fluorobenzyl)sulfanyl]aniline ≥95%

Supplier: Matrix Scientific

047573-500MG 047573-1G
101880-644EA 181.54 USD
101880-644 101880-646
4-[(4-Fluorobenzyl)sulfanyl]aniline ≥95%
4-[(4-Fluorobenzyl)sulfanyl]aniline

Hazard: Irritant

Formula: C₁₃H₁₂FNS
MDL Number: MFCD03551681
CAS Number: 710965-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR