3-Cyano-4-methylaniline ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-methylbenzonitrile

047570-5G 047570-25G
101880-634EA 23.08 USD
101880-634 101880-636
3-Cyano-4-methylaniline ≥95%
3-Cyano-4-methylaniline

Hazard: Irritant

Formula: C₈H₈N₂
MW: 132.16 g/mol
MDL Number: MFCD00017608
CAS Number: 50670-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR