2-[(4-Methylphenyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(4-Methylphenyl)thio]-5-nitrobenzaldehyde

047568-500MG 047568-1G
101880-626EA 181.54 USD
101880-626 101880-628
2-[(4-Methylphenyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%
2-[(4-Methylphenyl)sulfanyl]-5-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₁NO₃S
MW: 273.31 g/mol
MDL Number: MFCD00173708
CAS Number: 175278-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR