2-(Cyclohexylsulfanyl)-5-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Cyclohexylthio)-5-nitrobenzaldehyde

047567-500MG 047567-1G
101880-622EA 181.54 USD
101880-622 101880-624
2-(Cyclohexylsulfanyl)-5-nitrobenzenecarbaldehyde ≥95%
2-(Cyclohexylsulfanyl)-5-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₅NO₃S
MW: 265.33 g/mol
MDL Number: MFCD00084990
CAS Number: 175278-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR