2-[(4-Chlorophenyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitro-2-(4-chlorophenylthio)benzaldehyde

047564-500MG 047564-1G
101880-610EA 181.54 USD
101880-610 101880-612
2-[(4-Chlorophenyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%
2-[(4-Chlorophenyl)sulfanyl]-5-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₈ClNO₃S
MW: 293.73 g/mol
MDL Number: MFCD00173718
CAS Number: 42191-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR