2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

047553-500MG 047553-1G
101880-564EA 556.92 USD
101880-564 101880-566
2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile ≥95%
2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile

Hazard: Irritant

Formula: C₈H₄ClF₃N₂
MW: 220.58 g/mol
MDL Number: MFCD10568352
CAS Number: 13600-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR