1-(tert-Butyl)-2-methyl-4-bromo-1H-pyrrole-1,2-dicarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: 1-tert-Butyl 2-methyl 4-bromo-1H-pyrrole-1,2-dicarboxylate

047517-500MG 047517-5G
101880-412EA 355.38 USD
101880-412 101880-416
1-(tert-Butyl)-2-methyl-4-bromo-1H-pyrrole-1,2-dicarboxylate ≥97%
1-(tert-Butyl)-2-methyl-4-bromo-1H-pyrrole-1,2-dicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₄BrNO₄
MW: 304.15 g/mol
MDL Number: MFCD09751125
CAS Number: 156237-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR