N-Methoxy-N-methyl-2-thiophenecarboxamide ≥95%

Supplier: Matrix Scientific

047504-500MG 047504-5G
101880-350EA 84.62 USD
101880-350 101880-354
N-Methoxy-N-methyl-2-thiophenecarboxamide ≥95%
N-Methoxy-N-methyl-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₇H₉NO₂S
MDL Number: MFCD03643923
CAS Number: 229970-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR