5-Amino-1-(4-chlorobenzyl)-1H-1,2,3-triazole-4-carboxamide ≥95%

Supplier: Matrix Scientific

047445-500MG 047445-5G
101880-016EA 523.08 USD
101880-016 101880-020
5-Amino-1-(4-chlorobenzyl)-1H-1,2,3-triazole-4-carboxamide ≥95%
5-Amino-1-(4-chlorobenzyl)-1H-1,2,3-triazole-4-carboxamide

Hazard: Irritant

Formula: C₁₀H₁₀ClN₅O
MDL Number: MFCD03407124
CAS Number: 132269-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR