Ethyl-2,4-dichloronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2,4-dichloronicotinate

047418-500MG 047418-5G
101879-898EA 747.69 USD
101879-898 101879-902
Ethyl-2,4-dichloronicotinate ≥95%
Ethyl-2,4-dichloronicotinate

Hazard: Irritant

Formula: C₈H₇Cl₂NO₂
MW: 220.05 g/mol
MDL Number: MFCD12407388
CAS Number: 62022-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR