1-(4-Chlorophenyl)-2-oxo-3-pyrrolidinecarbonitrile ≥95%
Supplier: Matrix Scientific
047334-500MG
047334-5G
101879-554EA
278.46
USD
101879-554
101879-558
1-(4-Chlorophenyl)-2-oxo-3-pyrrolidinecarbonitrile ≥95%
1-(4-Chlorophenyl)-2-oxo-3-pyrrolidinecarbonitrile
Hazard: Irritant
Formula:
C₁₁H₉ClN₂O MW: 220.66 g/mol |
MDL Number:
MFCD09817456 CAS Number: 930298-99-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...