Benzyl-N-[cyclohexyl(phenylsulfonyl)methyl]carbamate ≥95%
Supplier: Matrix Scientific
047325-500MG
047325-5G
101879-506EA
181.54
USD
101879-506
101879-510
Benzyl-N-[cyclohexyl(phenylsulfonyl)methyl]carbamate ≥95%
Benzyl-N-[cyclohexyl(phenylsulfonyl)methyl]carbamate
Hazard: Irritant
Formula:
C₂₁H₂₅NO₄S MW: 387.5 g/mol |
MDL Number:
MFCD09972234 CAS Number: 232598-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...