2-(Dimethylamino)nicotinonitrile ≥95%

Supplier: Matrix Scientific

047255-500MG 047255-5G
101879-136EA 181.54 USD
101879-136 101879-140
2-(Dimethylamino)nicotinonitrile ≥95%
2-(Dimethylamino)nicotinonitrile

Hazard: Irritant

Formula: C₈H₉N₃
MDL Number: MFCD08689753
CAS Number: 60138-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR