Ethyl-4-(trifluoromethyl)-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

047195-500MG 047195-1G 047195-5G
101878-876EA 330.77 USD
101878-876 101878-878 101878-880
Ethyl-4-(trifluoromethyl)-2-pyridinecarboxylate ≥95%
Ethyl-4-(trifluoromethyl)-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD12498713
CAS Number: 1171919-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR