2-(Hydroxymethyl)-5-methoxy-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

047171-500MG 047171-1G
101878-762EA 750.77 USD
101878-762 101878-764
2-(Hydroxymethyl)-5-methoxy-4(1H)-pyridinone ≥95%
2-(Hydroxymethyl)-5-methoxy-4(1H)-pyridinone

Hazard: Irritant

Formula: C₇H₉NO₃
MDL Number: MFCD04122652
CAS Number: 6323-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR