Ethyl-2-(1H-pyrrolo[2,3-b]pyridin-1-yl)acetate ≥95%

Supplier: Matrix Scientific

047154-500MG 047154-1G
101878-696EA 375.38 USD
101878-696 101878-698
Ethyl-2-(1H-pyrrolo[2,3-b]pyridin-1-yl)acetate ≥95%
Ethyl-2-(1H-pyrrolo[2,3-b]pyridin-1-yl)acetate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD11100223
CAS Number: 1048913-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR