Methyl-2,4-dichloronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,4-dichloronicotinate

047150-500MG 047150-1G
101878-676EA 300 USD
101878-676 101878-678
Methyl-2,4-dichloronicotinate ≥95%
Methyl-2,4-dichloronicotinate

Hazard: Irritant

Formula: C₇H₅Cl₂NO₂
MW: 206.022 g/mol
MDL Number: MFCD11100222
CAS Number: 442903-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR