4-tert-Butyl-2-methyl-(2R,4R,5S)-5-phenylpyrrolidine-2,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

047111-500MG 047111-1G
101878-506EA 556.92 USD
101878-506 101878-508
4-tert-Butyl-2-methyl-(2R,4R,5S)-5-phenylpyrrolidine-2,4-dicarboxylate ≥95%
4-tert-Butyl-2-methyl-(2R,4R,5S)-5-phenylpyrrolidine-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₇H₂₃NO₄
MDL Number: MFCD12025934
CAS Number: 1219344-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR