2-Hydroxy-2-(4-propoxyphenyl)acetic acid ≥95%

Supplier: Matrix Scientific

047110-1G 047110-5G
101878-502EA 138.46 USD
101878-502 101878-504
2-Hydroxy-2-(4-propoxyphenyl)acetic acid ≥95%
2-Hydroxy-2-(4-propoxyphenyl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
MDL Number: MFCD07368282
CAS Number: 79694-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR