2-Amino-2-(3,4-dichlorophenyl)acetic acid ≥95%

Supplier: Matrix Scientific

047108-500MG 047108-1G
101878-494EA 446.15 USD
101878-494 101878-496
2-Amino-2-(3,4-dichlorophenyl)acetic acid ≥95%
2-Amino-2-(3,4-dichlorophenyl)acetic acid

Hazard: Irritant

Formula: C₈H₇Cl₂NO₂
MW: 220.06 g/mol
MDL Number: MFCD03425231
CAS Number: 128349-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR