2-Aminooxazole ≥95%

Supplier: Matrix Scientific

Synonyms: Oxazol-2-amine

047099-1G 047099-5G
101878-454EA 36.92 USD
101878-454 101878-456
2-Aminooxazole ≥95%
2-Aminooxazole

Hazard: Irritant

Formula: C₃H₄N₂O
MW: 84.10 g/mol
MDL Number: MFCD07364485
CAS Number: 4570-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR