2-Hydroxy-3-methoxybenzonitrile ≥95%

Supplier: Matrix Scientific

047096-500MG 047096-1G 047096-5G
101878-438EA 104.62 USD
101878-438 101878-440 101878-442
2-Hydroxy-3-methoxybenzonitrile ≥95%
2-Hydroxy-3-methoxybenzonitrile

Hazard: Irritant

Formula: C₈H₇NO₂
MDL Number: MFCD02261936
CAS Number: 6812-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR