tert-Butyl-4-[2-(3-formylphenoxy)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[2-(3-Formylphenoxy)ethyl]piperazine, N1-BOC protected

047055-500MG 047055-1G 047055-5G
101878-246EA 556.92 USD
101878-246 101878-248 101878-250
tert-Butyl-4-[2-(3-formylphenoxy)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-[2-(3-formylphenoxy)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₄
MW: 334.42 g/mol
MDL Number: MFCD08689721
CAS Number: 924869-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR