Ethyl-4-{2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-{2-hydroxy-2-[4-(methylsulphonyl)phenyl]ethyl}piperazine-1-carboxylate

047031-500MG 047031-1G 047031-5G
101878-126EA 278.46 USD
101878-126 101878-128 101878-130
Ethyl-4-{2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Ethyl-4-{2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₄N₂O₅S
MW: 356.44 g/mol
MDL Number: MFCD08689707
CAS Number: 924869-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR