2,4-Difluoro-5-methoxyaniline ≥95%
Supplier: Matrix Scientific
047019-500MG
047019-1G
101878-056EA
461.54
USD
101878-056
101878-058
2,4-Difluoro-5-methoxyaniline ≥95%
2,4-Difluoro-5-methoxyaniline
Hazard: Irritant
Formula:
C₇H₇NOF₂ |
MDL Number:
MFCD08689699 CAS Number: 98446-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...