Diethyl-2,3-pyridinedicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Diethyl pyridine-2,3-dicarboxylate

047016-500MG 047016-1G 047016-5G
101878-038EA 375.38 USD
101878-038 101878-040 101878-042
Diethyl-2,3-pyridinedicarboxylate ≥95%
Diethyl-2,3-pyridinedicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
MDL Number: MFCD08689697
CAS Number: 2050-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR