(2-Piperidinophenyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1-Piperidinyl)benzyl alcohol

046989-500MG 046989-1G 046989-5G
101877-920EA 258.46 USD
101877-920 101877-922 101877-924
(2-Piperidinophenyl)methanol ≥95%
(2-Piperidinophenyl)methanol

Hazard: Irritant

Formula: C₁₂H₁₇NO
MW: 191.28 g/mol
MDL Number: MFCD05864821
CAS Number: 87066-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR