Ethyl-1-oxaspiro[2.5]octane-6-carboxylate ≥92%

Supplier: Matrix Scientific

046953-1G 046953-5G
101877-782EA 183.08 USD
101877-782 102551-352
Ethyl-1-oxaspiro[2.5]octane-6-carboxylate ≥92%
Ethyl-1-oxaspiro[2.5]octane-6-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₆O₃
MW: 184.24 g/mol
MDL Number: MFCD12498695
CAS Number: 171361-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR